Labmaster Air-Drive Mixers
Use these mixers in the presence of volatile or explosive vapors, or where electrical power would be hazardous or impractical.
These mixers are excellent for pilot studies of chemical, food, and wastewater treatment processes. We offer LABMASTER air-drive mixers with two types of mounting styles. The tube suspension mixers require the heavy-duty clamp 04560-24 (sold separately). The universal clamp mixers mount directly to a ring stand or tank rim.
Mixers include:
— 5/16" dia x 12"L, 316 SS nonthrough-type shaft
— 5/16" bore x 2.5" dia, 316 SS A-310 impeller
— collet-type chuck
— universal clamp or tube suspension mounting
— muffler
— needle value
392,000.00 - ₹417,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Mounting Type Shaft Length (in)
Availability Pricing
Lightnin 869439 Air-drive mixer with tube suspension WW-50322-00 Mfr # 869439
COMPARE
1800 18
₹392,000.00INR / Each
Lightnin 869440 Air-drive mixer with universal clamp WW-50322-10 Mfr # 869440
COMPARE
1800 C-clamp 21.5
₹417,000.00INR / Each