Air lubricator, 1 to 20 scfm, 1/4" NPT(F)
Representative image only.
Air lubricator, 1 to 20 scfm, 1/4" NPT(F)
| Item: 0704270
₹18,600.00INR / Each
₹18,600.00INR / Each
Product Description
More About this Item
Air lubricator dispenses oil mist at less than 1 cfm. Uses light spindle oil. Porous bronze wick. Measures 4-1/4"L x 1-5/8" dia.
Specifications
  • Port Size1/4" NPT(F)
  • Thread Size1/4" NPT(F)
  • Power (Hz)NA
  • DescriptionAir lubricator, 1 to 20 scfm, 1/4" NPT(F)