LaMotte 1480 Turbidity Standard, 0 NTU, ISO/EPA, 60 ml Bottle

Representative image only.

LaMotte 1480 Turbidity Standard, 0 NTU, ISO/EPA, 60 ml Bottle

LaMotte | Mfr: 1480| Item: 0556525

Optimize water analysis

₹15,000.00INR / Each

₹15,000.00INR / Each

Buy It Together

Current Item

LaMotte 1480 Turbidity Standard, 0 NTU, ISO/EPA, 60 ml Bottle
LaMotte 1480 Turbidity Standard, 0 NTU, ISO/EPA, 60 ml Bottle
0556525

₹15,000.00INR / Each
+
LaMotte 0290-6 Turbidity Sample Test Tubes/Vials; 6/Pk
0556366

₹11,700.00INR / Pkg of 6
+
GFS Chemicals Turbidity Standard, 40.0 NTU, 1-L bottle
0839105

₹48,500.00INR / Each
+ + +
Hach 26597-49 StablCal <0.1 NTU Turbidity Standard, 500 mL
9951254

₹25,100.00INR / Each
=
Total Price 147900.00 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Choose from a variety of accessories

More About this Item

Replacement test kits, reagents, and standards for LaMotte products.

Specifications
  • DescriptionTurbidity Standard, 0 NTU, ISO/EPA, 60 ml Bottle
Documents & Certificates
Reviews
Q&A