LaMotte 5860 Dissolved Oxygen Test Kit, 50 tests/kit
Representative image only.
LaMotte 5860 Dissolved Oxygen Test Kit, 50 tests/kit
LaMotte | Mfr: 5860-01| Item: 5300300
Color titration method determines dissolved oxygen levels
Call For Price
Call For Price
Buy It Together
Current Item
LaMotte 5860 Dissolved Oxygen Test Kit, 50 tests/kit
LaMotte 5860 Dissolved Oxygen Test Kit, 50 tests/kit
5300300
Call For Price
+ + =
Total Price 3500.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Winkler titration method to test samples or calibrate meters
  • Inexpensive backup in case of meter failure
More About this Item
This test is an azide modification of the Winkler method where oxygen, in the presence of a strong alkali, oxidizes manganese, which in turn reacts with iodide to form iodine. This is titrated with a standard thiosulfate solution in the presence of a starch indicator to enhance the endpoint. Azide eliminates the nitrite interference. Test kit and refills contain enough reagents for 50 tests at an oxygen level of 10 ppm.
Specifications
  • TestDissolved oxygen
  • Measurement Points0 to 10 ppm
  • Tests Per Kit50
  • DescriptionDissolved Oxygen Test Kit, 50 tests/kit