GFS Chemicals Turbidity Standard, 40.0 NTU, 1-L bottle
Representative image only.
GFS Chemicals Turbidity Standard, 40.0 NTU, 1-L bottle
GFS Chemicals | Mfr: 85075| Item: 0839105
Ensure accurate readings by calibrating your turbidimeter
₹48,500.00INR / Each
₹48,500.00INR / Each
Buy It Together
Current Item
GFS Chemicals Turbidity Standard, 40.0 NTU, 1-L bottle
GFS Chemicals Turbidity Standard, 40.0 NTU, 1-L bottle
0839105
₹48,500.00INR / Each
+ + =
Total Price 99600.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • EPA-approved polymeric standards outlast Formazin standards
  • Suspended polymeric particles virtually never settle or clump
More About this Item
Primary Standard Solutions can be used to calibrate nephelometers or turbidimeters. Solutions are guaranteed to maintain NTU values within ±1% for one year. Available in one-liter bottles.
EPA United States Environmental protection agency
Specifications
  • NTU Value40
  • Volume (mL)1000
  • Volume (oz)33.8
  • DescriptionTurbidity Standard, 40.0 NTU, 1-L bottle