Labconco FlaskScrubber Upper Spindle Rack for FlaskScrubber and FlaskScrubber Vantage Washers
Representative image only.
Labconco FlaskScrubber Upper Spindle Rack for FlaskScrubber and FlaskScrubber Vantage Washers
Labconco | Mfr: 4595600| Item: 9896362
Optimize performance of your Labconco glassware washer
₹674,000.00INR / Each
₹674,000.00INR / Each
Buy It Together
Current Item
Labconco FlaskScrubber Upper Spindle Rack for FlaskScrubber and FlaskScrubber Vantage Washers
Labconco FlaskScrubber Upper Spindle Rack for FlaskScrubber and FlaskScrubber Vantage Washers
9896362
₹674,000.00INR / Each
+ +
SP Hotpack DI Water Boost Pump for Glassware Washer
7890362
₹228,000.00INR / Each
+ + + =
Total Price 10675000.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Adds more cleaning options to your washer
  • 1 year warranty
More About this Item
Choose from a wide range of rack accessories and other products to ensure top performance of your Labconco glassware washer.
Upper spindle rack provides injection cleaning and forced air drying in the FlaskScrubber and FlaskScrubber Vantage glassware washers. Includes side rails for installation. Rack includes side release latches, 30 removable spindles, 30 adjustable height clips, 10 small spindles, 15 glassware holders, spindle removal tool, 30 nylon plugs, and wash arm. Easily remove spindles and plug holes for accommodation of racks and inserts.
1 Year Warranty
Specifications
  • DescriptionUpper Spindle Rack for FlaskScrubber and FlaskScrubber Vantage Washers