InnoCal NIST-Traceable Calibration; Pipette, 16 Channel
Representative image only.
InnoCal NIST-Traceable Calibration; Pipette, 16 Channel
InnoCal | Item: 1710404
Ensure your instrument remains precise
₹221,000.00INR / Each
₹221,000.00INR / Each
Buy It Together
Current Item
InnoCal NIST-Traceable Calibration; Pipette, 16 Channel
InnoCal NIST-Traceable Calibration; Pipette, 16 Channel
1710404
₹221,000.00INR / Each
+ + + +
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109
₹8,400.00INR / Each
+ + =
Total Price 625600.00 ADD 7 ITEMS TO CART
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Confirm that your item meets the manufacturer's published specification
  • Meet EPA, USDA, ISO, and GLP/GMP standards
  • NIST-Traceable calibration for 16 channel pipettors
More About this Item
InnoCal is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA) under certificate number 1746.01. This accreditation is your assurance that the calibration is performed in a qualified, controlled laboratory.
Calibrations on pipettes, dispensers and graduated cylinders are performed per the procedures in ISO8655 using a precision pipette accuracy tester. Before testing, the valves and seals on pipettes and dispensers are tested for integrity using a leak tester. Variable pipettors and dispensers are measured at 3 points at 10, 50 and 100% of the maximum value. Fixed pipettors and dispensers are measured at the nominal value. Evaluation of the unit under test is done for dispensed volume and the coefficient of variability (CV) value.
This service is for 16 channel pipettors.
Specifications
  • Calibration TypeLiquid Testing
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points3 points, 10 aliquots per channel; 16 channel
  • Calibration Test Points20, 50 and 100% of full scale
  • Calibration Test Rangeup to 4000 mL
  • DescriptionNIST-Traceable Calibration, Pipette 16 Channel