InnoCal A2LA ISO17025 Accredited Calibration; Pipette, 16 Channel
Representative image only.
InnoCal A2LA ISO17025 Accredited Calibration; Pipette, 16 Channel
InnoCal | Item: 1750115
Ensure your instrument remains precise
₹228,000.00INR / Each
₹228,000.00INR / Each
Buy It Together
Current Item
InnoCal A2LA ISO17025 Accredited Calibration; Pipette, 16 Channel
InnoCal A2LA ISO17025 Accredited Calibration; Pipette, 16 Channel
1750115
₹228,000.00INR / Each
+ + + + + +
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109
₹8,400.00INR / Each
=
Total Price 632600.00 ADD 7 ITEMS TO CART
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Confirm that your item meets the manufacturer's published specification
  • Meet EPA, USDA, ISO, and GLP/GMP standards
  • A2LA Accredited Calibration. View our scope of accreditation
  • NIST-Traceable calibration for 16 channel pipettors
More About this Item
InnoCal is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA) under certificate number 1746.01. This accreditation is your assurance that the calibration is performed in a qualified, controlled laboratory.
Specifications
  • Calibration TypeA2LA ISO17025 Accredited Liquid Testing
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points3 points, 10 aliquots per channel; 16 channel
  • Calibration Test Points20, 50 and 100% of full scale
  • Calibration Test Rangeup to 4000 mL
  • DescriptionA2LA ISO17025 Accredited Calibration; Pipette, 16 Channel