IKA 3378000 ETS-D5 Programmable Temperature Probe
Representative image only.
IKA 3378000 ETS-D5 Programmable Temperature Probe
IKA | Mfr: 3378000| Item: 0467155
₹31,761.00INR / Each
for an additional
₹30,000.00INR / Each
for an additional
₹42,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹31,761.00INR / Each
Buy It Together
Current Item
IKA 3378000 ETS-D5 Programmable Temperature Probe
IKA 3378000 ETS-D5 Programmable Temperature Probe
0467155
₹31,761.00INR / Each
+ + +
InnoCal NIST-Traceable Calibration; Digital Indicator
1710161
₹30,000.00INR / Each
=
Total Price 237461.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Sensor features optimized PID control with three modes of operation — ensuring perfect temperature control without overshooting the set temperature, even in the case of quick heating. Mode A is suitable for work with varying parameters and is safety temperature adjustable; Mode B is suitable for series operation under uniform conditions; Mode C is suitable for unsupervised operation.
Two year warranty
Specifications
  • DescriptionETS-D5 Programmable Temperature Probe