IKA Ceramic-Top Hot Plates
Precise digital control
  • Exact temperature setting via digital display
  • Extremely fast heating
The flat white ceramic top plate cleans easily, and resists alkalies and acids. This microprocessor-controlled hot plate features an over-temperature safety circuit fixed at 1022°F (550°C).
Order optional temperature sensor 04671-55 for more precise temperature control. The sensor features optimized PID control with three modes of operation — ensuring perfect temperature control without overshooting the set temperature, even in the case of quick heating. Mode A is suitable for work with varying parameters and is safety temperature adjustable; Mode B is suitable for series operation under uniform conditions; Mode C is suitable for unsupervised operation.
66,851.00 - ₹315,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Length (in)
Availability Pricing
IKA 3581601 C-MAG HP 4 THERM Glass-Ceramic Hot Plate, 4" x 4"; 115 VAC 0467160 Mfr # 3581601
COMPARE
115 4
₹172,000.00INR / Each
IKA 3581600 C-MAG HP 4 THERM Glass-Ceramic Hot Plate, 4" x 4"; 230 VAC 0467162 Mfr # 3581600
COMPARE
230 4
₹66,851.00INR / Each
IKA 3581801 C-MAG HP 7 THERM Glass-Ceramic Hot Plate, 7" x 7"; 115 VAC 0467164 Mfr # 3581801
COMPARE
115 7
₹236,000.00INR / Each
IKA 3581800 C-MAG HP 7 THERM Glass-Ceramic Hot Plate, 7" x 7"; 230 VAC 0467166 Mfr # 3581800
COMPARE
230 7
₹78,109.00INR / Each
IKA 3582001 C-MAG HP 10 THERM Glass-Ceramic Hot Plate, 10" x 10"; 115 VAC 0467168 Mfr # 3582001
COMPARE
115 10
₹315,000.00INR / Each
IKA 3582000 C-MAG HP 10 THERM Glass-Ceramic Hot Plate, 10" x 10"; 230 VAC 0467170 Mfr # 3582000
COMPARE
230 10
₹99,808.00INR / Each