IKA 3159000 Laboratory Mixer Vessel Strap Clamp with Boss Head Clamp
Representative image only.
IKA 3159000 Laboratory Mixer Vessel Strap Clamp with Boss Head Clamp
IKA | Mfr: 3159000| Item: 5070371
Get the most out of your IKA Mixer with these accessories
₹53,100.00INR / Each
₹53,100.00INR / Each
Buy It Together
Current Item
IKA 3159000 Laboratory Mixer Vessel Strap Clamp with Boss Head Clamp
IKA 3159000 Laboratory Mixer Vessel Strap Clamp with Boss Head Clamp
5070371
₹53,100.00INR / Each
+ + + + +
IKA Eurostar 20 Digital Mixer, 30 to 2000 rpm, 115 VAC
5070230
₹504,000.00INR / Each
+ + +
Cole-Parmer Heavy-Duty Stand for Overhead Mixer
9996821
₹202,000.00INR / Each
=
Total Price 2893787.00 ADD 9 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • For vessel diameters of 1-1/2" to 11-3/4" (40-300 mm).
  • 2 year warranty
More About this Item
A variety of impellers, paddles, clamps, and stands are available.
For securing vessels against walls or for synchronized rotation during stirring or dispersing. Includes boss head clamp 50703-97. Use with stands 50703-93 or 50703-94.
Two year warranty
Specifications
  • DescriptionLaboratory Mixer Vessel Strap Clamp with Boss Head Clamp