IKA Dispersing Elements for T-25 Digital High-Speed Homogenizer Systems
Handle sample volumes from 1 to 2000 mL
  • Ideal for use with tissue and other fibrous materials
  • Suitable for abrasive substances
  • aAppropriate for solvents
Select a generator according to your sample volume, working solution, and needed dispersion type. The sawtooth generator is available in four sample volume ranges with 10um or 15um ultimate fineness options.
The homogenizer, stand, and clamps are sold separately.
157,168.00 - ₹210,448.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Tip Diameter (mm)
Availability Pricing
IKA Sawtooth Dispersing Element for T-25 Homogenizer, 10 G ST 0473792 Mfr # 0004447100
COMPARE
10
₹157,168.00INR / Each
IKA Sawtooth Dispersing Element for T-25 Homogenizer, 18 G ST 0473794 Mfr # 0004447300
COMPARE
18
₹206,051.00INR / Each
IKA Sawtooth Dispersing Element for T-25 Homogenizer, 25 G ST 0473796 Mfr # 0004447500
COMPARE
25
₹210,448.00INR / Each