IKA 3008600 Laboratory Mixer Vessel Strap Clamp
Representative image only.
IKA 3008600 Laboratory Mixer Vessel Strap Clamp
IKA | Mfr: 3008600| Item: 5070370
Get the most out of your IKA Mixer with these accessories
₹12,438.00INR / Each
₹12,438.00INR / Each
Buy It Together
Current Item
IKA 3008600 Laboratory Mixer Vessel Strap Clamp
IKA 3008600 Laboratory Mixer Vessel Strap Clamp
5070370
₹12,438.00INR / Each
+ + +
Cole-Parmer Heavy-Duty Stand for Overhead Mixer
9996821
₹202,000.00INR / Each
+ + +
IKA Eurostar 20 Digital Mixer, 30 to 2000 rpm, 115 VAC
5070230
₹504,000.00INR / Each
+ + =
Total Price 2853125.00 ADD 9 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
A variety of impellers, paddles, clamps, and stands are available.
For securing vessels against walls or for synchronized rotation during stirring or dispersing. Use with stands 50703-90, 50703-91, or 50703-92. For vessel diameters of 1 1/2'' to 11 3/4'' (40-300 mm).
Two year warranty
Specifications
  • DescriptionLaboratory Mixer Vessel Strap Clamp