Idex Threaded Luer Adapter, Red ETFE, 0.050" Bore, Female Luer x Female 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Threaded Luer Adapter, Red ETFE, 0.050" Bore, Female Luer x Female 1/4-28 Flat Bottom; 1/EA
Idex | Mfr: P-678| Item: 0201422
Convenience of quick connect and disconnect of a luer connection
₹2,700.00INR / Each
₹2,700.00INR / Each
Buy It Together
Current Item
Product Description
Key Features
  • Manufactured from inert polymers
  • For use with Cole-Parmer BT-T, BA-T, PT-T, and PA-T Touchscreen Series Balances
  • 1 month warranty
  • 1 month warranty
More About this Item
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
1 Month Warranty
Specifications
  • MaterialRed ETFE
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Female 1/4-28 Flat Bottom
  • DescriptionThreaded Luer Adapter, Red ETFE, 0.050" Bore, Female Luer x Female 1/4-28 Flat Bottom; 1/EA