Idex Threaded Luer Adapter, Natural Polypropylene, 0.040" Bore, Female Luer x Female 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Threaded Luer Adapter, Natural Polypropylene, 0.040" Bore, Female Luer x Female 1/4-28 Flat Bottom; 1/EA
Idex | Mfr: P-628P| Item: 0202342
Convenience of quick connect and disconnect of a luer connection
₹1,900.00INR / Each
₹1,900.00INR / Each
Buy It Together
Current Item
Product Description
Key Features
  • Manufactured from inert polymers
  • Female Luer x Female 1/4-28 Flat Bottom
  • 0.040" thru-hole diameter
  • Manufactured from natural polypropylene
  • 1 year warranty
More About this Item
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
1 Year Warranty
Specifications
  • MaterialNatural Polypropylene
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Female 1/4-28 Flat Bottom
  • DescriptionThreaded Luer Adapter, Natural Polypropylene, 0.040" Bore, Female Luer x Female 1/4-28 Flat Bottom; 1/EA