Idex Threaded Luer Adapter, Natural ETFE, 0.050" Bore, Female Luer x Male 10-32 Coned; 1/EA
Representative image only.
Idex Threaded Luer Adapter, Natural ETFE, 0.050" Bore, Female Luer x Male 10-32 Coned; 1/EA
Idex | Mfr: P-642| Item: 0201406
Convenience of quick connect and disconnect of a luer connection
₹1,500.00INR / Each
₹1,500.00INR / Each
Buy It Together
Current Item
Product Description
Key Features
  • Manufactured from inert polymers
  • Density determination kit for analytical balances
  • 1 month warranty
More About this Item
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
1 Month Warranty
Specifications
  • MaterialNatural ETFE
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Male 10-32 Coned
  • DescriptionThreaded Luer Adapter, Natural ETFE, 0.050" Bore, Female Luer x Male 10-32 Coned; 1/EA