Idex Threaded Luer Adapter, Black PEEK, 0.050" Bore, Female Luer x Female M6 Flat Bottom; 1/EA
Representative image only.
Idex Threaded Luer Adapter, Black PEEK, 0.050" Bore, Female Luer x Female M6 Flat Bottom; 1/EA
Idex | Mfr: P-660| Item: 0202359
Convenience of quick connect and disconnect of a luer connection
₹4,800.00INR / Each
₹4,800.00INR / Each
Buy It Together
Current Item
Product Description
Key Features
  • Manufactured from inert polymers
  • Female Luer x Female M6 Flat Bottom
  • 0.050" thru-hole diameter
  • Manufactured from Black PEEK
  • 1 year warranty
More About this Item
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
1 Year Warranty
Specifications
  • MaterialBlack PEEK
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Female M6 Flat Bottom
  • DescriptionThreaded Luer Adapter, Black PEEK, 0.050" Bore, Female Luer x Female M6 Flat Bottom; 1/EA