Idex Threaded Luer Adapter, Black ETFE, 0.050" Bore, Female Luer x Male M6 Flat Bottom; 1/EA
Representative image only.
Idex Threaded Luer Adapter, Black ETFE, 0.050" Bore, Female Luer x Male M6 Flat Bottom; 1/EA
Idex | Mfr: P-686| Item: 0201425
Convenience of quick connect and disconnect of a luer connection
₹1,500.00INR / Each
₹1,500.00INR / Each
Buy It Together
Current Item
Product Description
Key Features
  • Manufactured from inert polymers
  • Works with with Cole-Parmer MB-T-21 Microbalance 10100-28
  • 1 month warranty
More About this Item
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
1 Month Warranty
Specifications
  • MaterialBlack ETFE
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Male M6 Flat Bottom
  • DescriptionThreaded Luer Adapter, Black ETFE, 0.050" Bore, Female Luer x Male M6 Flat Bottom; 1/EA