Idex SuperFlangeless™ Female Nut, Standard Knurl, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex SuperFlangeless™ Female Nut, Standard Knurl, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Idex | Mfr: P-420| Item: 0202317
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
₹2,500.00INR / Each
₹2,500.00INR / Each
Buy It Together
Current Item
Idex SuperFlangeless™ Female Nut, Standard Knurl, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Idex SuperFlangeless™ Female Nut, Standard Knurl, Natural PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
0202317
₹2,500.00INR / Each
+ + + + + + =
Total Price 48910.00 ADD 7 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Female nut
  • Natural PEEK
  • 1 year warranty
More About this Item
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection Type1/4-28 Flat Bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.588
  • ColorNatural
  • Max Operating Pressure (PSI)1000
  • DescriptionSuperFlangeless™ Nut, Female, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA