Idex Manual Selection Valve, Analytical, SS/Vespel®, 6-Position, 7-Port, 1/16" OD Tubing; 1/EA
Representative image only.
Idex Manual Selection Valve, Analytical, SS/Vespel®, 6-Position, 7-Port, 1/16" OD Tubing; 1/EA
Idex | Mfr: 7060| Item: 0201524
Allow operator control of low-use application valves
₹294,000.00INR / Each
₹294,000.00INR / Each
Buy It Together
Current Item
Idex Manual Selection Valve, Analytical, SS/Vespel®, 6-Position, 7-Port, 1/16" OD Tubing; 1/EA
Idex Manual Selection Valve, Analytical, SS/Vespel®, 6-Position, 7-Port, 1/16" OD Tubing; 1/EA
0201524
₹294,000.00INR / Each
+ +
Idex Stator for Valve Models 7060/7066, SS; 1/EA
0201555
₹131,000.00INR / Each
=
Total Price 804000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Enable connections among multiple fluid systems 6-position, 7-port configuration
  • 10-32 coned ports for 1/16" OD tubing
  • 1 year warranty
More About this Item
Manual valves serve well in applications that require operator control or involve lesser frequency of use. Selection valves enable connection to one column for another without the need to manually disconnect the plumbing. In sequencing or diagnostic applications, the valve alternates between the sample or reagent streams.
1 Year Warranty
Specifications
  • Outlet Port10-32 coned
  • Inlet Port10-32 coned
  • Max Pressure (PSI)7000
  • Max Pressure (bar)483
  • Wetted MaterialsSS/Vespel
  • Tubing OD (in)0.0625
  • DescriptionManual Selection Valve, 6-Way, SS/Vespel®, 6-Position, 7-Port, 1/16" OD Tubing; 1/EA