Idex Stator for Valve Models 7060/7066, SS; 1/EA
Representative image only.
Idex Stator for Valve Models 7060/7066, SS; 1/EA
Idex | Mfr: 7060-039| Item: 0201555
Prolong the life of your valves
₹131,000.00INR / Each
₹131,000.00INR / Each
Product Description
Key Features
  • Stators need replacement only if the ports or sealing surfaces become damaged
  • Use with valve models 7060/7066
  • 1 year warranty
More About this Item
Optimize performance of your manual chromatography valves with these replacement stators.
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithValve models 7060/7066
  • DescriptionStator for Valve Models 7060/7066, SS; 1/EA