Idex Barbed Connector, Y, Polypropylene, 3/32" Hose Barb; 1/EA
Representative image only.
Idex Barbed Connector, Y, Polypropylene, 3/32" Hose Barb; 1/EA
Idex | Mfr: P-861| Item: 0202408
Make solid connections in low-pressure applications
₹880.00INR / Each
₹880.00INR / Each
Buy It Together
Current Item
Idex Barbed Connector, Y, Polypropylene, 3/32" Hose Barb; 1/EA
Idex Barbed Connector, Y, Polypropylene, 3/32" Hose Barb; 1/EA
0202408
₹880.00INR / Each
+ + + =
Total Price 10280.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Offers less turbulence and gentler mixing than Tee connectors
  • Manufactured from polypropylene
  • For use with 3/32" ID soft-walled tubing
  • 1 year warranty
More About this Item
Use barbed Y connectors to firmly join together three pieces of soft-walled peristaltic tubing with similar inner diameters.
1 Year Warranty
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeHPLC/Chromatography
  • Barb Size / Tubing Id (Port 1) (mm)2.4
  • Barb Size / Tubing Id (Port 1) (in)3/32
  • Barb Size / Tubing Id (Port 2) (mm)2.4
  • Barb Size / Tubing Id (Port 2) (in)3/32
  • Barb Size / Tubing Id (Port 3) (mm)2.4
  • Barb Size / Tubing Id (Port 3) (in)3/32
  • ColorNatural
  • DescriptionLow-Pressure Connector, Barbed Y, Polypropylene, 3/32" ID Tubing/Barb; 1/EA