Cole-Parmer VapLock™ Barbed Y Fitting, Polypropylene, 1/4" ID Tubing; 10/PK
Representative image only.
Cole-Parmer VapLock™ Barbed Y Fitting, Polypropylene, 1/4" ID Tubing; 10/PK
Cole-Parmer | Item: 1202088
Connect soft tubing using barbed fittings
₹4,900.00INR / Each
₹4,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Barbed Y Fitting, Polypropylene, 1/4" ID Tubing; 10/PK
Cole-Parmer VapLock™ Barbed Y Fitting, Polypropylene, 1/4" ID Tubing; 10/PK
1202088
₹4,900.00INR / Each
+ + + =
Total Price 11780.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Y fitting for 1/4" ID tubing
More About this Item
These Y-connector barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Specifications
  • Fitting MaterialPolypropylene
  • Number Of Ports3
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • Barb Size / Tubing Id (Port 2) (in)1/4
  • Barb Size / Tubing Id (Port 3) (in)1/4
  • ColorTranslucent
  • DescriptionBarbed Y Fitting, Polypropylene, 1/4" ID Tubing; 10/PK