Cole-Parmer® Cleanroom Hose Barb Fittings, Y Union, Animal-Derivative-Free Polypropylene
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily combine or split fluid flow using Y unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on each port prevents tubing from rotating, minimizing kinks. These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
1,240.00 - ₹3,920.00INR / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
COMPARE
3/32 3/32 3/32
₹1,260.00INR / Pkg of 10
COMPARE
5/8 5/8 5/8
₹3,160.00INR / Pkg of 10
COMPARE
3/4 3/4 3/4
₹3,200.00INR / Pkg of 10
COMPARE
1 1 1
₹3,920.00INR / Pkg of 10
COMPARE
1/16 1/16 1/16
₹1,260.00INR / Pkg of 10
COMPARE
5/32 5/32 5/32
₹1,900.00INR / Pkg of 10
COMPARE
1/8 1/8 1/8
₹1,240.00INR / Pkg of 10
COMPARE
3/16 3/16 3/16
₹1,900.00INR / Pkg of 10
COMPARE
1/4 1/4 1/4
₹2,250.00INR / Pkg of 10
COMPARE
5/16 5/16 5/16
₹2,270.00INR / Pkg of 10
COMPARE
5/8 5/8 5/8
₹2,270.00INR / Pkg of 10
COMPARE
1/2 1/2 1/2
₹2,400.00INR / Pkg of 10
Showing 1- 12 of 12