Cole-Parmer Hose Barb Fitting, Y Union, Animal-Derivative-Free Polypropylene, Cleanroom Packed, 5/16" ID; 10/Pk
Representative image only.
Cole-Parmer Hose Barb Fitting, Y Union, Animal-Derivative-Free Polypropylene, Cleanroom Packed, 5/16" ID; 10/Pk
Cole-Parmer | Item: 5011145
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹4,300.00INR / Pkg of 10
₹4,300.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer Hose Barb Fitting, Y Union, Animal-Derivative-Free Polypropylene, Cleanroom Packed, 5/16" ID; 10/Pk
Cole-Parmer Hose Barb Fitting, Y Union, Animal-Derivative-Free Polypropylene, Cleanroom Packed, 5/16" ID; 10/Pk
5011145
₹4,300.00INR / Pkg of 10
+ + + + + + + + + =
Total Price 21150.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
  • 5/16" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily combine or split fluid flow using Y unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on each port prevents tubing from rotating, minimizing kinks. These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolypropylene
  • Sterilization MethodGamma irradiation between 25 and 50 kGy, EtO, or autoclave up to 121°C (250°F)
  • Connection TypeHose Barb Y
  • Number Of Ports3
  • Barb Size / Tubing Id (Port 1) (in)5/16
  • Barb Size / Tubing Id (Port 2) (in)5/16
  • Barb Size / Tubing Id (Port 3) (in)5/16
  • Flow PatternY
  • ColorNatural
  • Min Temperature (° F)-9
  • Max Temperature (° F)150
  • Min Temperature (° C)-23
  • Max Temperature (° C)66
  • Regulatory ApprovalsNon-DEHP;RoHS;ADCF;FDA;Non-Phthalate
  • DescriptionHose Barb Fitting, Y Union, Animal-Derivative-Free Polypropylene, Cleanroom Packed, 5/16" ID; 10/Pk