Cole-Parmer Hose Barb Fitting, Y Union, HDPE, Cleanroom Packed, 3/16" ID; 10/Pk
Representative image only.
Cole-Parmer Hose Barb Fitting, Y Union, HDPE, Cleanroom Packed, 3/16" ID; 10/Pk
Cole-Parmer | Item: 5011136
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹3,900.00INR / Pkg of 10
₹3,900.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer Hose Barb Fitting, Y Union, HDPE, Cleanroom Packed, 3/16" ID; 10/Pk
Cole-Parmer Hose Barb Fitting, Y Union, HDPE, Cleanroom Packed, 3/16" ID; 10/Pk
5011136
₹3,900.00INR / Pkg of 10
+ =
Total Price 8800.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520 (when used unmodified for the manufacture of food contact articles) and RoHS compliant
  • 3/16" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily combine or split fluid flow using Y unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on each port prevents tubing from rotating, minimizing kinks. These high-density polyethylene (HDPE) fittings are softer than nylon, polypropylene, and Kynar®. They exhibit excellent resistance to chemicals but have lower thermal resistance than other materials.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialHDPE
  • Connection TypeHose Barb Y
  • Number Of Ports3
  • Barb Size / Tubing Id (Port 1) (in)3/16
  • Barb Size / Tubing Id (Port 2) (in)3/16
  • Barb Size / Tubing Id (Port 3) (in)3/16
  • Flow PatternY
  • ColorWhite
  • Regulatory ApprovalsFDA;RoHS;Non-DEHP;Non-Phthalate
  • DescriptionHose Barb Fitting, Y Union, HDPE, Cleanroom Packed, 3/16" ID; 10/Pk