Humboldt H-6225 Accuflame Straight Tube Gas TIRrill Bunsen Burner, Lp Gas
Representative image only.
Humboldt H-6225 Accuflame Straight Tube Gas TIRrill Bunsen Burner, Lp Gas
Humboldt | Mfr: H-6225| Item: 3613012
₹10,400.00INR / Each
₹10,400.00INR / Each
Buy It Together
Current Item
Humboldt H-6225 Accuflame Straight Tube Gas TIRrill Bunsen Burner, Lp Gas
Humboldt H-6225 Accuflame Straight Tube Gas TIRrill Bunsen Burner, Lp Gas
3613012
₹10,400.00INR / Each
+ =
Total Price 19600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 3 month warranty
More About this Item
Straight Tube Gas Burners feature adjustable flames for very slow heating or quick boiling. Flame retainer prevents flame lifting or extinguishing. Tapered serrated gas inlet connections for easy tube
mounting.
3 Month Warranty
Specifications
  • Gas TypeLiquid Propane (LP)
  • BTU Output (per hour)3200
  • Inlet Tube OD (in)0.4375
  • Height (in)6
  • DescriptionAccuflame Straight Tube Gas TIRrill Bunsen Burner, Lp Gas