Straight Tube Bunsen Burners with Tirrill Bunner
Straight Tube Gas Burners feature adjustable flames for very slow heating or quick boiling. Flame retainer prevents flame lifting or extinguishing. Tapered serrated gas inlet connections for easy tube
mounting.
10,400.00 - ₹25,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
BTU Output (per hour) Gas Type Inlet Tube OD (in)
Availability Pricing
Humboldt H-6220 Accuflame Bunsen Burner, TIRrill Burner, Natural Gas 3613010 Mfr # H-6220
COMPARE
1200 Natural Gas 0.4375
₹25,200.00INR / Each
Humboldt H-6225 Accuflame Straight Tube Gas TIRrill Bunsen Burner, Lp Gas 3613012 Mfr # H-6225
COMPARE
3200 Liquid Propane (LP) 0.4375
₹10,400.00INR / Each