Humboldt H-2087 Accuflame Gas Line for Bunsen Burners, Flexible Steel Covered with Rubber/Cloth, 36" L
Representative image only.
Humboldt H-2087 Accuflame Gas Line for Bunsen Burners, Flexible Steel Covered with Rubber/Cloth, 36" L
Humboldt | Mfr: H-2087| Item: 3613070
Optimize performance of your Bunsen burner with this durable gas line
₹9,200.00INR / Each
₹9,200.00INR / Each
Buy It Together
Current Item
Humboldt H-2087 Accuflame Gas Line for Bunsen Burners, Flexible Steel Covered with Rubber/Cloth, 36" L
Humboldt H-2087 Accuflame Gas Line for Bunsen Burners, Flexible Steel Covered with Rubber/Cloth, 36" L
3613070
₹9,200.00INR / Each
+ + =
Total Price 37200.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of durable steel covered with rubber and cloth
  • 3 month warranty
More About this Item
Ensure optimal usage of your Bunsen burner with this flexible steel gas line. This gas line is 36” (91.4 cm) long with a 1/4” ID. At each of the line, there is a rubber connector with 3/8” ID, for quick and easy connection.
3 Month Warranty
Specifications
  • DescriptionAccuflame Gas Line for Bunsen Burners, Flexible Steel Covered with Rubber/Cloth, 36" L