Humboldt H-5550 Accuflame Bunsen Burner, High-Temperature Burner, Natural Gas
Representative image only.
Humboldt H-5550 Accuflame Bunsen Burner, High-Temperature Burner, Natural Gas
Humboldt | Mfr: H-5550| Item: 3613020
₹18,300.00INR / Each
₹18,300.00INR / Each
Buy It Together
Current Item
Humboldt H-5550 Accuflame Bunsen Burner, High-Temperature Burner, Natural Gas
Humboldt H-5550 Accuflame Bunsen Burner, High-Temperature Burner, Natural Gas
3613020
₹18,300.00INR / Each
+ =
Total Price 27500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 3 month warranty
More About this Item
Gas Burner distributes flame through 100 separate openings in stainless steel grid top for uniform heating. The grid diameter is 11⁄2" (3.8 cm). This burner gives twice the heat of a straight tube burner. This burner features a venturi tube for thorough mixing of gas and air, and adjustable gas orifice and air shutter for complete combustion. Adjustable flame size. Rustproof chrome-plated brass body with zinc-aluminum alloy base.
3 Month Warranty
Specifications
  • Gas TypeNatural Gas
  • BTU Output (per hour)1200
  • Inlet Tube OD (in)0.4375
  • Height (in)8 1/2
  • DescriptionAccuflame Bunsen Burner, High-Temperature Burner, Natural Gas
  • Warranty3 Months