Honeywell Chart Paper, Thermal, 12", Blank with No Range or Time

Representative image only.

Honeywell Chart Paper, Thermal, 12", Blank with No Range or Time

Honeywell | Mfr: 30755317-001| Item: 8066390

Replacement paper for your Honeywell chart recorder

₹19,500.00INR / Pkg of 100

₹19,500.00INR / Pkg of 100

Buy It Together

Current Item

Honeywell Chart Paper, Thermal, 12", Blank with No Range or Time
Honeywell Chart Paper, Thermal, 12", Blank with No Range or Time
8066390

₹19,500.00INR / Pkg of 100
+ +
Honeywell Replacement Chart Recorder Thermal Pen
8066392

₹138,000.00INR / Each
=
Total Price 1426500.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Varying temperature ranges and speeds
  • 18 month warranty

More About this Item

Choose the paper compatible with your chart recorder and application needs.

18 Month Warranty
Specifications
  • Chart TypeThermal
  • Chart SpeedBlank
  • Chart Diameter (in)12
  • DescriptionChart Paper, Thermal, 12", Blank with No Range or Time
  • Warranty18 months
Reviews
Q&A