Honeywell Chart Paper, 10", 0-600, 24 Hours

Representative image only.

Honeywell Chart Paper, 10", 0-600, 24 Hours

Honeywell | Mfr: 24001660-003| Item: 2604673

Replacement paper for your Honeywell chart recorder

₹14,200.00INR / Each

₹14,200.00INR / Each

Buy It Together

Current Item

Honeywell Chart Paper, 10", 0-600, 24 Hours
Honeywell Chart Paper, 10", 0-600, 24 Hours
2604673

₹14,200.00INR / Each
+ +
Honeywell Replacement Chart Recorder Thermal Pen
8066392

₹138,000.00INR / Each
=
Total Price 1421200.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Varying temperature ranges and speeds
  • 1 year warranty

More About this Item

Choose the paper compatible with your chart recorder and application needs.

1 Year Warranty
Specifications
  • Chart TypeCircular
  • Chart Speed24 Hours
  • Chart Diameter (in)10
  • DescriptionChart Paper, 10", 0-600, 24 Hours
  • Warranty1 year
Reviews
Q&A