Extech SDL200 4-Channel Datalogging Thermometer with Sd Card Memory
Representative image only.
Extech SDL200 4-Channel Datalogging Thermometer with Sd Card Memory
Extech | Mfr: SDL200| Item: 9500132
Records data onto an SD card in Excel® format for easy transfer to a PC for analysis
₹123,000.00INR / Each
for an additional
₹39,900.00INR / Each
for an additional
₹29,100.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹123,000.00INR / Each
Buy It Together
Current Item
Extech SDL200 4-Channel Datalogging Thermometer with Sd Card Memory
Extech SDL200 4-Channel Datalogging Thermometer with Sd Card Memory
9500132
₹123,000.00INR / Each
+ + + =
Total Price 233300.00 ADD 4 ITEMS TO CART
Product Description
Includes
type K beaded wire temperature probe, 2 GB SD card, USB cable, carrying case, and six AA batteries.
Key Features
  • Data logging capability up to 20,000 records using a 2G SD card
  • User programmable sampling rate from 1 to 3600 seconds
  • Manual store and recall of up to 99 records
  • Records readings with real-time/date stamp
  • 1 year warranty
More About this Item
Four-channel data logging thermometer accepts both thermocouple and RTD probes. Data is recorded onto a standard SD memory card, instead of a built-in memory; the advantage being that when the memory is full the SD card can be simply changed out for unlimited data storage.
The removable SD card is easy to transport and insert into a computer’s memory card slot or SD card reader for upload. The data is provided in Excel, allowing you to easily analyze the information without the need for additional software. Data can also be streamed directly to a computer using the RS-232 port on the meter.
Features include min/max and hold functions, temperature differential, offset adjustment, date-and-time stamp, auto power off and low-battery indication.
Needs USB adapter 93956-04.
1 Year Warranty
Specifications
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels4
  • Types Of Probes AcceptedType E, Type J, Type K, Type R, Type S, Type T
  • Min Temperature (° F)-328
  • Max Temperature (° F)2501
  • Min Temperature (° C)-200
  • Max Temperature (° C)1372
  • AccuracyType E, J, K, T: ±0.4% of reading +1.8°F (+1.0°C); Type R, S: ±0.5% of reading +2.0°F (+1.0°C)
  • Resolution0.1°
  • OutputSD card
  • Data LoggingYes
  • Memory Capacity20000 data points
  • No. of Probe(s) Included1
  • Battery6 AA (included)
  • Description4-Channel Datalogging Thermometer with Sd Card Memory