Extech PH100 Exstik Waterproof Pocket pH Tester

Representative image only.

Extech PH100 Exstik Waterproof Pocket pH Tester

Extech | Mfr: PH100| Item: 5302654

Flat-surface electrode is great for semisolids, solids, and surface testing

Call For Price

for an additional
₹18,700.00INR / Each
for an additional
₹25,500.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Extech PH100 Exstik Waterproof Pocket pH Tester
Extech PH100 Exstik Waterproof Pocket pH Tester
5302654

Call For Price
+
Extech PH105 ExStik Flat Surface Electrode
5302656

₹14,700.00INR / Each
+
InnoCal NIST-Traceable Calibration; pH Tester or Pen
1710145

₹18,700.00INR / Each
=
Total Price 33400.00 ADD 3 ITEMS TO CART
Product Description

Includes

sensor cap, sample cup, neck strap, and four CR2032 batteries.

Key Features

  • Built-in memory recalls up to 15 labeled readings
  • Smart chip tell you when to calibrate or replace your meter
  • 1 year warranty

More About this Item

ExStik pH tester features a flat pH sensor making it ideal for testing semisolids and surfaces. Flat surface also reduces the chance of accidental breakage of the electrode tip—perfect for rugged field use! Built-in smart chip simplifies calibration and error diagnosis. CAL alert notifies you when it is time to recalibrate ensuring accurate readings. Renew indicator tells you when it is time to replace the electrode. Built-in memory recalls up to 15 sequentially tagged readings. LCD simultaneously shows pH and temperature readings, plus an analog bar graph of pH values lets you view reading trends. Waterproof tester floats in water.

1 Year Warranty
Specifications
  • Meter TypeTester
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min Temperature (° C)-5
  • Max Temperature (° C)90
  • Min Temperature (° F)23
  • Max Temperature (° F)194
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery4 CR2032 (included)
  • Width (cm)3.4925
  • Height (cm)4.1275
  • Width (in)1 3/8
  • Height (in)1 5/8
  • DescriptionExstik Waterproof Pocket pH Tester
  • Warranty1 year
Reviews
Q&A