InnoCal A2LA ISO17025 Accredited Calibration; pH Tester or Pen
Representative image only.
InnoCal A2LA ISO17025 Accredited Calibration; pH Tester or Pen
InnoCal | Item: 1750097
Services that ensure the accuracy of your pH Measurement instrumentation
₹25,500.00INR / Each
₹25,500.00INR / Each
Buy It Together
Current Item
InnoCal A2LA ISO17025 Accredited Calibration; pH Tester or Pen
InnoCal A2LA ISO17025 Accredited Calibration; pH Tester or Pen
1750097
₹25,500.00INR / Each
+
Traceable Calibrated Portable Conductivity Meter
1960103
₹72,600.00INR / Each
+ + + + + + + + =
Total Price 884300.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • A2LA Accredited Calibration. View our scope of accreditation
  • 1 year warranty
  • Up to three calibration points against NIST-traceable pH buffers
  • 1 year warranty
More About this Item
Regulatory and quality agencies like FDA and EPA have strict guidelines for the determination and control of pH. Maintain your compliance with the help of InnoCal. We can calibrate new instrument purchases from Cole-Parmer, recalibrate your existing instruments, and provide maintenance and repair services.
InnoCal’s metrologists can service most any brand of pH, conductivity, or dissolved oxygen meter including Oakton Instruments, accumet, Fisher Brand, Mettler-Toledo, YSI, WTW, Horiba, Hanna Instruments, Thermo Scientific Orion, VWR SympHony, and others.
Specifications
  • Calibration TypeA2LA ISO17025 Accredited Liquid Testing
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points3 pH Points, 1 Temperature Point
  • Calibration Test Points4, 7, 10 pH values, 25 C
  • Calibration Test Range4 to 10 pH
  • DescriptionA2LA ISO17025 Accredited Calibration; pH Tester or Pen