Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/4" NPT(M) x 5/16" ID; 1/Each
Representative image only.
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/4" NPT(M) x 5/16" ID; 1/Each
Eldon James | Mfr: A4-5SS| Item: 5012463
Threaded barb adapter design provides safe, difficult-to-break connection
₹3,600.00INR / Each
₹3,600.00INR / Each
Buy It Together
Current Item
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/4" NPT(M) x 5/16" ID; 1/Each
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/4" NPT(M) x 5/16" ID; 1/Each
5012463
₹3,600.00INR / Each
+ + + + +
Alicat Mass Flowmeter with Display, 0 - 500 SCCM
1530880
₹169,000.00INR / Each
+
Alicat Mass Flow Controller with Display, 0 - 200 SCCM
1530902
₹222,000.00INR / Each
+
Alicat Liquid Flow Controller with Display, 0 - 5 CCM
1530931
₹258,000.00INR / Each
+
Alicat Power Adapter, 24 VDC, 1 Amp (Worldwide)
1530939
₹4,800.00INR / Each
+ =
Total Price 1167300.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
  • 1/4" NPT(M) x 5/16" hose barb
More About this Item
Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place.
These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialStainless Steel
  • Sterilization MethodE-beam/gamma irradiation, EtO, or autoclave up to 134°C
  • Number Of Ports2
  • Connection TypeThreaded NPT(M) x Hose Barb
  • Thread Size NPT(M) (Port 1) (InchShort)1/4
  • Barb Size / Tubing Id (Port 2) (in)5/16
  • Flow PatternStraight
  • ColorMetallic
  • Regulatory ApprovalsNon-Phthalate;RoHS;Non-DEHP
  • DescriptionThreaded to Hose Barb Fitting, Straight Adapter, Stainless Steel, 1/4" NPT(M) x 5/16" ID