Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 1/8" ID; 10/PK
Representative image only.
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 1/8" ID; 10/PK
Eldon James | Mfr: A4-2NN| Item: 5012413
Threaded barb adapter design provides safe, difficult-to-break connection
₹1,800.00INR / Pkg of 10
₹1,800.00INR / Pkg of 10
Buy It Together
Current Item
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 1/8" ID; 10/PK
Eldon James Threaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 1/8" ID; 10/PK
5012413
₹1,800.00INR / Pkg of 10
+ + + + + + + + + =
Total Price 29700.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
  • 1/4" NPT(M) x 1/8" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks.
These natural nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialNylon
  • Sterilization MethodE-beam/gamma irradiation, EtO, or autoclave up to 121°C
  • Connection TypeThreaded NPT(M) x Hose Barb
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/4
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Flow PatternStraight
  • ColorWhite
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;FDA;RoHS
  • DescriptionThreaded to Hose Barb Fitting, Straight Adapter, Natural Nylon, 1/4" NPT(M) x 1/8" ID; 10/PK