Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • Biopharma compatible and USP 661
  • BPOG compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. The elbows feature a 90° bend and is designed to fit in a tight space. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kink.
These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
5,100.00INR / Pkg of 10
47 variations of this product are available.
See All Product Options Below
47 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Flow Pattern Connection Type
Availability Pricing
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®  1/8" NPT x 3/32" ID; 10/PK 5012689 Mfr # L2-1.5-200NK7
COMPARE
3/32 90° elbow Threaded NPT(M) x Hose Barb
₹5,100.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 3/4" NPT x 5/8" ID; 10/PK 5012814 Mfr # L12-10NK7
COMPARE
5/8 90° elbow Threaded NPT(M) x Hose Barb
₹8,900.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 3/4" NPT x 3/4" ID; 10/PK 5012815 Mfr # L12-12NK7
COMPARE
3/4 90° elbow Threaded NPT(M) x Hose Barb
₹8,900.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 3/4" NPT x 1" ID; 10/PK 5012816 Mfr # L12-16NK7
COMPARE
1 90° elbow Threaded NPT(M) x Hose Barb
₹9,900.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 3/4" NPT x 1/4" ID; 10/PK 5012817 Mfr # L12-4NK7
COMPARE
1/4 90° elbow Threaded NPT(M) x Hose Barb
₹7,200.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 3/4" NPT x 3/8" ID; 10/PK 5012818 Mfr # L12-6NK7
COMPARE
3/8 90° elbow Threaded NPT(M) x Hose Barb
₹7,300.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 3/4" NPT x 1/2" ID; 10/PK 5012819 Mfr # L12-8NK7
COMPARE
1/2 90° elbow Threaded NPT(M) x Hose Barb
₹7,500.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1" NPT x 3/4" ID; 10/PK 5012820 Mfr # L16-12NK7
COMPARE
3/4 90° elbow Threaded NPT(M) x Hose Barb
₹10,700.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1" NPT x 1" ID; 10/PK 5012821 Mfr # L16-16NK7
COMPARE
1 90° elbow Threaded NPT(M) x Hose Barb
₹10,700.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/8" NPT x 1/16" ID; 10/PK 5012822 Mfr # L2-1-200NK7
COMPARE
1/16 90° elbow Threaded NPT(M) x Hose Barb
₹5,100.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/8" NPT x 5/32" ID; 10/PK 5012823 Mfr # L2-2.5NK7
COMPARE
5/32 90° elbow Threaded NPT(M) x Hose Barb
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/8" NPT x 1/8" ID; 10/PK 5012824 Mfr # L2-2NK7
COMPARE
1/8 90° elbow Threaded NPT(M) x Hose Barb
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/8" NPT x 3/16" ID; 10/PK 5012825 Mfr # L2-3NK7
COMPARE
3/16 90° elbow Threaded NPT(M) x Hose Barb
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/8" NPT x 1/4" ID; 10/PK 5012826 Mfr # L2-4NK7
COMPARE
1/4 90° elbow Threaded NPT(M) x Hose Barb
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/8" NPT x 5/16" ID; 10/PK 5012827 Mfr # L2-5NK7
COMPARE
5/16 90° elbow Threaded NPT(M) x Hose Barb
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/8" NPT x 3/8" ID; 10/PK 5012828 Mfr # L2-6NK7
COMPARE
3/8 90° elbow Threaded NPT(M) x Hose Barb
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/8" BSPT x 5/32" ID; 10/PK 5012829 Mfr # L2B-2.5NK7
COMPARE
5/32 90° elbow Threaded BSPT(M) x Hose Barb
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/8" BSPT x 1/8" ID; 10/PK 5012830 Mfr # L2B-2NK7
COMPARE
1/8 90° elbow Threaded BSPT(M) x Hose Barb
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/8" BSPT x 1/4" ID; 10/PK 5012831 Mfr # L2B-4NK7
COMPARE
1/4 90° elbow Threaded BSPT(M) x Hose Barb
₹4,600.00INR / Pkg of 10
Eldon James Threaded to Hose Barb Fittings, 90° Elbow, Natural Kynar®, 1/4" NPT x 1/4" ID; 10/PK 5012837 Mfr # L4-4NK7
COMPARE
1/4 90° elbow Threaded NPT(M) x Hose Barb
₹5,000.00INR / Pkg of 10
Showing 1 - 20 of 47