Cole-Parmer® Cleanroom NPT(M) to Hose Barb Fittings, Straight Adapter, Natural Kynar®
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • Biopharma compatible and USP 661 and BPOG compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks. These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
1,650.00 - ₹7,940.00INR / Pkg of 10
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Thread size NPT(M) (port 1) (in)
Availability Pricing
COMPARE
3/4 Natural Kynar® 3/4
₹5,270.00INR / Pkg of 10
COMPARE
1 Natural Kynar® 3/4
₹7,940.00INR / Pkg of 10
COMPARE
1/4 Natural Kynar® 3/4
₹4,430.00INR / Pkg of 10
COMPARE
3/8 Natural Kynar® 3/4
₹4,540.00INR / Pkg of 10
COMPARE
1/2 Natural Kynar® 3/4
₹4,650.00INR / Pkg of 10
COMPARE
1 Natural Kynar® 1
₹6,050.00INR / Pkg of 10
COMPARE
1/8 Natural Kynar® 1/8
₹1,650.00INR / Pkg of 10
COMPARE
3/16 Natural Kynar® 1/8
₹2,340.00INR / Pkg of 10
COMPARE
1/4 Natural Kynar® 1/8
₹2,340.00INR / Pkg of 10
COMPARE
3/8 Natural Kynar® 1/8
₹2,750.00INR / Pkg of 10
COMPARE
1/4 Natural Kynar® 1/4
₹2,880.00INR / Pkg of 10
COMPARE
3/8 Natural Kynar® 1/4
₹3,050.00INR / Pkg of 10
COMPARE
1/2 Natural Kynar® 1/4
₹3,160.00INR / Pkg of 10
COMPARE
1/4 Natural Kynar® 3/8
₹3,240.00INR / Pkg of 10
COMPARE
5/16 Natural Kynar® 3/8
₹3,320.00INR / Pkg of 10
COMPARE
3/8 Natural Kynar® 3/8
₹3,380.00INR / Pkg of 10
COMPARE
1/2 Natural Kynar® 3/8
₹2,920.00INR / Pkg of 10
COMPARE
3/4 Natural Kynar® 1/2
₹4,270.00INR / Pkg of 10
COMPARE
1/4 Natural Kynar® 1/2
₹3,590.00INR / Pkg of 10
COMPARE
3/8 Natural Kynar® 1/2
₹3,730.00INR / Pkg of 10
Showing 1 - 20 of 21