Digi-Sense Temperature Probe, Type J, 6" L, 1/8" Dia, PTFE, Mini-Connector
Representative image only.
Digi-Sense Temperature Probe, Type J, 6" L, 1/8" Dia, PTFE, Mini-Connector
Digi-Sense | Item: 9044631
Davis Instruments can provide many additional configurations beyond those listed here
₹15,300.00INR / Each
for an additional
₹30,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹15,300.00INR / Each
Buy It Together
Current Item
Digi-Sense Temperature Probe, Type J, 6" L, 1/8" Dia, PTFE, Mini-Connector
Digi-Sense Temperature Probe, Type J, 6" L, 1/8" Dia, PTFE, Mini-Connector
9044631
₹15,300.00INR / Each
+ =
Total Price 58000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 316 stainless steel sheath
  • 1 year warranty
More About this Item
Magnesium Oxide insulated thermocouples, commonly referred to as MgO thermocouples, are used in many process and laboratory applications. Many desirable characteristics make them an excellent choice for general and special purpose applications. They are available in all thermocouple element types, single or dual elements, a wide variety of sheath diameters / materials, rugged in nature, bendable, and have fairly high temperature ratings. The thermocouples listed are commonly used; however, Davis Inotek Instruments can provide many additional configurations beyond those listed.
1 Year Warranty
Specifications
  • Probe TypeJ
  • Connector TypeMini ANSI
  • Sheath Length (in)6
  • Sheath Diameter (InchShort)0.125
  • DescriptionTemperature Probe, Type J, 6" L, 1/8" Dia, PTFE, Mini-Connector