Diba Omnifit® T-Series Solvent Bottle Cap, GL45, 3 Luer Ports and 1 Venting Port, Blue
Representative image only.
Diba Omnifit® T-Series Solvent Bottle Cap, GL45, 3 Luer Ports and 1 Venting Port, Blue
Diba | Mfr: 00945T-3W| Item: 2194319
Connect a vapor trap using the vent port to prevent solvent vapors from escaping
₹7,900.00INR / Each
₹7,900.00INR / Each
Buy It Together
Current Item
Diba Omnifit® T-Series Solvent Bottle Cap, GL45, 3 Luer Ports and 1 Venting Port, Blue
Diba Omnifit® T-Series Solvent Bottle Cap, GL45, 3 Luer Ports and 1 Venting Port, Blue
2194319
₹7,900.00INR / Each
+ + =
Total Price 67200.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Use without any fittings or use male Luer fittings to connect various tubing sizes
  • Anti-twist design
More About this Item
The T-series solvent bottle caps help contain vapors and protect lab staff from exposure to toxic fumes. Caps feature a 1/4"-28 UNF(F) vent port for connection to a vapor trap, allowing pressure equalization and prevention of solvent vapor escape. Push through 1/8" OD semi-rigid tubing for a liquid- and vapor-tight seal or use standard male Luer fittings to connect other tubing sizes. Luer port lets you also connect a syringe for liquid removal. Anti-twist cap design allows the PTFE body to spin freely within the outer cap. No more twisted tubes and no need to disconnect fluid lines before unscrewing the cap. All caps are autoclavable.
Use activated carbon vapor traps with 1/4"-28 UNF(M) connection to prevent any dangerous solvent from escaping into the laboratory. Traps feature an indicator strip that changes color when saturated. Connect 1/16" or 1/8" ID flexible tubing or 1/16” OD semi-rigid tubing to caps with Luer adapters. Bottle thread adapter converts bottles with S40 threads to accept any GL45 thread caps. Plug any unused ports with a Luer plug to ensure a closed system.
Specifications
  • Cap MaterialPTFE inner body/PP outer cap
  • Cap / Stopper SizeGL45
  • Cap ColorBlue
  • Special Features3 x Luer
  • AutoclavableYes
  • DescriptionDiba Omnifit® T-Series Solvent Waste Cap, GL45, 3 Luer ports and 1 venting port, blue; 1/ea