Diba Omnifit® T-Series Solvent Waste Caps with Vent
Connect a vapor trap using the vent port to prevent solvent vapors from escaping
  • Use without any fittings or use male Luer fittings to connect various tubing sizes
  • Anti-twist design
The T-series solvent bottle caps help contain vapors and protect lab staff from exposure to toxic fumes. Caps feature a 1/4"-28 UNF(F) vent port for connection to a vapor trap, allowing pressure equalization and prevention of solvent vapor escape. Push through 1/8" OD semi-rigid tubing for a liquid- and vapor-tight seal or use standard male Luer fittings to connect other tubing sizes. Luer port lets you also connect a syringe for liquid removal. Anti-twist cap design allows the PTFE body to spin freely within the outer cap. No more twisted tubes and no need to disconnect fluid lines before unscrewing the cap. All caps are autoclavable.
Use activated carbon vapor traps with 1/4"-28 UNF(M) connection to prevent any dangerous solvent from escaping into the laboratory. Traps feature an indicator strip that changes color when saturated. Connect 1/16" or 1/8" ID flexible tubing or 1/16” OD semi-rigid tubing to caps with Luer adapters. Bottle thread adapter converts bottles with S40 threads to accept any GL45 thread caps. Plug any unused ports with a Luer plug to ensure a closed system.
7,700.00 - ₹50,300.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Special Features
Availability Pricing
Diba Omnifit® T-Series Solvent Bottle Cap, GL38/38-430 (glass), 3 Luer Ports and 1 Venting Port, Blue 2194294 Mfr # 00938T-3W
COMPARE
38-430 (GL38) 3 x Luer
₹8,000.00INR / Each
Diba Omnifit® T-Series Solvent Bottle Cap, GL38/38-430 (glass), 4 Luer Ports and 1 Venting Port, Blue 2194297 Mfr # 00938T-4W
COMPARE
38-430 (GL38) 4 x Luer
₹7,700.00INR / Each
Diba Omnifit® T-Series Solvent Bottle Cap, GL45, 3 Luer Ports and 1 Venting Port, Blue 2194319 Mfr # 00945T-3W
COMPARE
GL45 3 x Luer
₹7,900.00INR / Each
Diba Omnifit® T-Series Solvent Bottle Cap, GL45, 4 Luer Ports and 1 Venting Port, Blue 2194323 Mfr # 00945T-4W
COMPARE
GL45 4 x Luer
₹13,400.00INR / Each
Diba Omnifit® T-Series Solvent Bottle Cap, 53B, 4 Luer Ports and 1 Venting Port, Blue 2194324 Mfr # 00953T-4W
COMPARE
53B 4 x Luer
₹20,100.00INR / Each
Diba Omnifit® T-Series Solvent Bottle Cap, 83B, 4 Luer Ports and 1 Venting Port, Blue 2194325 Mfr # 00983T-4W
COMPARE
83B 4 x Luer
₹49,000.00INR / Each
Diba Omnifit® T-Series Solvent Bottle Cap, 83B, 8 Luer Ports and 1 Venting Port, White 2194326 Mfr # 00983T-8W
COMPARE
83B 8 x Luer
₹50,300.00INR / Each
Showing 1- 7 of 7