Diba Omnifit® Activated Carbon Vapor Trap for Solvent Waste Caps with Venting Port; 1/ea
Representative image only.
Diba Omnifit® Activated Carbon Vapor Trap for Solvent Waste Caps with Venting Port; 1/ea
Diba | Mfr: 009CF-TW| Item: 2194330
Prevent chemical vapors from escaping from solvent containers
₹57,600.00INR / Each
₹57,600.00INR / Each
Product Description
Key Features
  • Compatible with Diba T-series solvent caps
  • Connects to 1/4"-28 UNF vent port
More About this Item
Use traps with Luer connection with T-series solvent waste caps without valve and filter. Traps with 1/4"-28 UNF(M) connection works with caps with vent. Traps with UNF connection feature an indicator strip that changes color as it gets saturated.
Use activated carbon trap with T-series solvent caps with vent port. An indicator strip changes color to show when the vapor trap is saturated.
Specifications
  • Compatible WithT-series solvent caps with vent; 1/4"-28 UNF(M) connection