Delmhorst BD-10/PKG General Contractor Moisture Meter with Electrode
Representative image only.
Delmhorst BD-10/PKG General Contractor Moisture Meter with Electrode
Delmhorst | Mfr: BD-10/PKG| Item: 5982039
Great entry level meter
₹138,000.00INR / Each
₹138,000.00INR / Each
Buy It Together
Current Item
Delmhorst BD-10/PKG General Contractor Moisture Meter with Electrode
Delmhorst BD-10/PKG General Contractor Moisture Meter with Electrode
5982039
₹138,000.00INR / Each
+ =
Total Price 165500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Easy-to-read analog display
  • Wood Scale - 6% - 40% moisture range for flooring and building material such as wood studs, floor joists, subfloors
  • Reference Scale - Reads from 0 - 100 on a relative basis on non-wood materials such as concrete, plaster, and insulation.
  • 1 year warranty
More About this Item
Easy to operate and easy to read, the BD-10 measures moisture in wood and other non-wood building materials. For those who prefer an analog display, the BD-10 is a great alternative to the digital BD-2100. The BD-10 is a great entry level meter, and is ready to go with a single button press!
1 Year Warranty
Specifications
  • Measurement TypePin
  • Battery1 9V (included)
  • DescriptionGeneral Contractor Moisture Meter with Electrode