Delmhorst 26-ES Moisture Meter Electrode, Hammer Type
Representative image only.
Delmhorst 26-ES Moisture Meter Electrode, Hammer Type
Delmhorst | Mfr: 26-ES| Item: 5982050
Configure your meter to your needs
₹37,200.00INR / Each
₹37,200.00INR / Each
Buy It Together
Current Item
Delmhorst 26-ES Moisture Meter Electrode, Hammer Type
Delmhorst 26-ES Moisture Meter Electrode, Hammer Type
5982050
₹37,200.00INR / Each
+ +
Delmhorst FX-2000W/PS Digital Hay Moisture Meter
5982316
₹132,000.00INR / Each
+
Delmhorst PC/KIT Moisture Meter Computer Interface Kit
5982045
₹47,000.00INR / Each
=
Total Price 310800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Flexibility to choose the right combination of electrodes and accessories for your application
  • 3 month warranty
More About this Item
Delmhorst offers the widest selections of electrodes and accessories in the industry. This allows you to customize you meter to meet your specific requirement, and each electrode is interchangeable with any Delmhorst moisture meter with a quick connection..
Designed so that the pins can be driven into hardened materials. The electrode is also insulated with only the tips being exposed, providing the only way to measure moisture content at various depths of penetration. Supplied with 2” pins for 1-1/4” penetration (pin 496).
3 Month Warranty
Specifications
  • Measurement TypeHammer
  • DescriptionMoisture Meter Electrode, Hammer Type