Delmhorst F-6 Analog Hay Moisture Meters

Prevent hay spoilage and fire hazards
  • Color-coded range at optimum moisture content shows at a glance if hay is safe to bale
  • Simplified controls are user friendly for entry-level use
  • Built-in calibration check ensures accurate readings
This hay moisture meter is ideal for agricultural applications where you need to know if your crop is ready for baling and safe storage. Color-coding the optimum range of 18 to 22% MC (moisture content) lets you know at a glance if you need to let the hay dry further or if it needs to be wetted before baling.
Available in two ranges to suit requirements for both US domestic (13 to 40% MC) and export (6 to 30% MC) markets.
80,200.00 - ₹125,000.00INR / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Moisture Range Non-Wood Scale (Max) (%) Moisture Range Non-Wood Scale (Min) (%) Description
Availability Pricing
Delmhorst F-6/6-30 F-6/6-30 Analog Hay Moisture Meter Only, Range 6% - 30% 5982313 Mfr # F-6/6-30
COMPARE
40 13 F-6/6-30 Analog Hay Moisture Meter Only, Range 6% - 30%
₹80,200.00INR / Each
Delmhorst F-6/6-30/1235 F-6/6-30 Analog Hay Moisture Meter, Range 6% - 30% with 10" Probe 5982314 Mfr # F-6/6-30/1235
COMPARE
30 6 F-6/6-30 Analog Hay Moisture Meter, Range 6% - 30% with 10" Probe
₹94,600.00INR / Each
Delmhorst F-6/6-30/PKG F-6/6-30 Analog Hay Moisture Meter Kit, Range 6% - 30% 5982315 Mfr # F-6/6-30/PKG
COMPARE
30 6 F-6/6-30 Analog Hay Moisture Meter Kit, Range 6% - 30%
₹125,000.00INR / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat