Delmhorst FX-2000 Digital Hay Moisture Meter
Protect crop value and prevent dangerous conditions
  • Dual-mode programming allows spot checks or continuous monitoring
  • Connect power supply to 12 V battery to save power while in continuous mode
  • Store up to 100 readings with average and maximum
  • Audible alarm sounds when the moisture reading exceeds the set point
This digital hay moisture meter continuously measures moisture content during the baling process using the bale chamber sensor and a 12 V battery, such as those used in tractors and trucks. The meter is also compatible with bale prods to monitor the bale curing process.
Built-in calibration check ensures readings are accurate and backlight provides visibility in low-light conditions.
132,000.00 - ₹207,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Delmhorst FX-2000W/PS Digital Hay Moisture Meter 5982316 Mfr # FX-2000W/PS
COMPARE
Digital Hay Moisture Meter
₹132,000.00INR / Each
Delmhorst FX-2000/PKG Digital Hay Moisture Meter Package 5982318 Mfr # FX-2000/PKG
COMPARE
Digital Hay Moisture Meter Package
₹207,000.00INR / Each