Delmhorst 22-E General Purpose Electrode 3-1/4" Penetration
Representative image only.
Delmhorst 22-E General Purpose Electrode 3-1/4" Penetration
Delmhorst | Mfr: 22-E| Item: 5982048
Configure your meter to your needs
₹22,800.00INR / Each
₹22,800.00INR / Each
Buy It Together
Current Item
Delmhorst 22-E General Purpose Electrode 3-1/4" Penetration
Delmhorst 22-E General Purpose Electrode 3-1/4" Penetration
5982048
₹22,800.00INR / Each
+
Delmhorst PC/KIT Moisture Meter Computer Interface Kit
5982045
₹47,000.00INR / Each
+ +
Delmhorst FX-2000W/PS Digital Hay Moisture Meter
5982316
₹132,000.00INR / Each
=
Total Price 296400.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Flexibility to choose the right combination of electrodes and accessories for your application
  • 3 month warranty
More About this Item
Delmhorst offers the widest selections of electrodes and accessories in the industry. This allows you to customize you meter to meet your specific requirement, and each electrode is interchangeable with any Delmhorst moisture meter with a quick connection..
Use to measure moisture on material up to 1-1/2” thick, such as wood, drywall, plaster and masonry. Supplied with 1” pins for 1/2” penetration (pin 2497/A-100).
3 Month Warranty
Specifications
  • Measurement TypePin
  • DescriptionGeneral Purpose Electrode 3-1/4" Penetration