Delmhorst 830-4 36" Bale Prod - Requires H-4 Handle (59823-50)

Representative image only.

Delmhorst 830-4 36" Bale Prod - Requires H-4 Handle (59823-50)

Delmhorst | Mfr: 830-4| Item: 5982322

Configure your meter to your needs

₹22,100.00INR / Each

₹22,100.00INR / Each

Buy It Together

Current Item

Delmhorst 830-4 36" Bale Prod - Requires H-4 Handle (59823-50)
Delmhorst 830-4 36" Bale Prod - Requires H-4 Handle (59823-50)
5982322

₹22,100.00INR / Each
+ +
Delmhorst FX-2000W/PS Digital Hay Moisture Meter
5982316

₹132,000.00INR / Each
+
Delmhorst PC/KIT Moisture Meter Computer Interface Kit
5982045

₹47,000.00INR / Each
=
Total Price 295700.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Flexibility to choose the right combination of electrodes and accessories for your application
  • 1 year warranty

More About this Item

Delmhorst offers the widest selections of electrodes and accessories in the industry. This allows you to customize you meter to meet your specific requirement, and each electrode is interchangeable with any Delmhorst moisture meter with a quick connection..

1 Year Warranty
Specifications
  • Measurement TypePin
  • Description36" Bale Prod - Requires H-4 Handle (59823-50)
  • Warranty1 year
Reviews
Q&A