Delmhorst 43-E Moisture Meter Electrode, Clamp-Type
Representative image only.
Delmhorst 43-E Moisture Meter Electrode, Clamp-Type
Delmhorst | Mfr: 43-E| Item: 5982011
Configure your meter to your needs
₹80,200.00INR / Each
₹80,200.00INR / Each
Buy It Together
Current Item
Delmhorst 43-E Moisture Meter Electrode, Clamp-Type
Delmhorst 43-E Moisture Meter Electrode, Clamp-Type
5982011
₹80,200.00INR / Each
+
Delmhorst PC/KIT Moisture Meter Computer Interface Kit
5982045
₹47,000.00INR / Each
+ +
Delmhorst FX-2000W/PS Digital Hay Moisture Meter
5982316
₹132,000.00INR / Each
=
Total Price 339400.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Flexibility to choose the right combination of electrodes and accessories for your application
  • 3 month warranty
More About this Item
Delmhorst offers the widest selections of electrodes and accessories in the industry. This allows you to customize you meter to meet your specific requirement, and each electrode is interchangeable with any Delmhorst moisture meter with a quick connection..
The clamp-type electrode is used to measure moisture in flat paper stock, fiberboard and veneer
3 Month Warranty
Specifications
  • Measurement TypeClamp
  • DescriptionMoisture Meter Electrode, Clamp-Type